MilliporeSigma
  • The effect of Piper laetispicum extract (EAE-P) during chronic unpredictable mild stress based on interrelationship of inflammatory cytokines, apoptosis cytokines and neurotrophin in the hippocampus.
The effect of Piper laetispicum extract (EAE-P) during chronic unpredictable mild stress based on interrelationship of inflammatory cytokines, apoptosis cytokines and neurotrophin in the hippocampus.
BMC complementary and alternative medicine (2015-07-18)
Hui Xie, Di Jin, Yun Kang, Xueru Shi, Hongrui Liu, Haixing Shen, Jian Chen, Macheng Yan, Juan Liu, ShengLi Pan
ABSTRACT
The Piper laetispicum C.DC. (Piperaceae) is a traditionally used herb in China for invigorating circulation and reducing stasis, detumescence and analgesia, which is distributed in the southern part of China and the southeastern part of Asia. Previous studies demonstrated that the ethyl acetate extract (EAE-P) of P. laetispicum possesses a significant antidepressant-like effect at doses higher than 60 mg/kg in Kunming (KM) mice, and this effect was not due to an increase in locomotive activity. To research this mechanism, in the present study, the chronic unpredictable mild stress (CUMS) model in Sprague-Dawley rats was used to further elucidate behavioral changes and corresponding changes in inflammatory cytokines (TNF-α, IL-6, IL-10), apoptosis cytokines (P53, Bax, Bcl2, caspase-3) and neurotrophin (BDNF) in the hippocampus of EAE-P treatment animals. The results suggest that EAE-P is beneficial to the behavioral outcome of the CUMS model animals, and decreased amounts of inflammatory cytokine IL-6 contributed to the antidepressant-like activation of EAE-P in every dosage group (15, 30, 60 mg/kg). In the low dosage group, down-regulated apoptosis cytokine p53 is associated with EAE-P effect, but it is inflammatory cytokine TNF-α that is related to the effect of EAE-P in the high dosage group. Meanwhile, the P53-dependent antiapoptotic effect of EAE-P may not be through Bcl-2 and Bax modulation. Furthermore, EAE-P showed up-regulated expression of brain-derived neurotrophic factor (BDNF) mRNA and down-regulated apoptosis cytokine caspase-3 mRNA, which was the same change tendency as with Fluoxetine.
Product Description
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethyl acetate, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethyl acetate, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, JIS 1000, ≥99.5%, suitable for residue analysis