MilliporeSigma
  • Identification of a highly sulfated fucoidan from sea cucumber Pearsonothuria graeffei with well-repeated tetrasaccharides units.
Identification of a highly sulfated fucoidan from sea cucumber Pearsonothuria graeffei with well-repeated tetrasaccharides units.
Carbohydrate polymers (2015-10-03)
Yaqin Hu, Shan Li, Junhui Li, Xingqian Ye, Tian Ding, Donghong Liu, Jianchu Chen, Zhiwei Ge, Shiguo Chen
ABSTRACT
Sea cucumber fucoidan is a major bioactive component of sea cucumber. The structures of fucoidans have significant influences on their biological activities. The present study clarified the delicate structure of a fucoidan from Pearsonothuria graeffei. Fucoidan was obtained after papain digestion and purified by ion chromatography. The carbohydrate sequence of fucoidan was firstly determined by negative-ion electrospray tandem mass spectrometry (ES-MS) with collision-induced dissociation of the oligosaccharide fragments, which were obtained by mild acid hydrolysis, and completed by NMR for assignment of the anomeric conformation. It was unambiguously identified as a tetrasaccharide repeating unit with a backbone of [ → 3Fuc (2S, 4S) α1 → 3Fucα1→ 3Fuc (4S) α1 → 3Fuc#7 × 10#]n. The glycosidic bonds between the non-sulfated and 2,4-O-disulfated fucose residues were selectively cleaved, and highly ordered oligosaccharide fragments with a tetrasaccharide repeating unit were obtained. The highly 4-O- and 2, 4-di-O-sulfated polysaccharide deserves further developments for Pharmacia use.
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence