MilliporeSigma
  • Simultaneous Quantification of Five Bioactive Flavonoids in High Altitude Plant Actinocarya tibetica by LC-ESI-MS/MS.
Simultaneous Quantification of Five Bioactive Flavonoids in High Altitude Plant Actinocarya tibetica by LC-ESI-MS/MS.
Journal of AOAC International (2015-08-14)
Baljinder Singh, Shreyans K Jain, Sandip B Bharate, Manoj Kushwaha, Ajai Prakash Gupta, Ram A Vishwakarma
ABSTRACT
An LC/MS method has been developed for the simultaneous quantification of five flavonoids, i.e., mosloflavone, negletein, gardenin B, 5-methoxy-6,7-methylenedioxyflavone, and 5,6,7-trimethoxyflavone, in different ultrasound assisted solvent extracts of Actinocarya tibetica. The chromatographic separation was achieved on a Chromolith Speed ROD RP-18e column with gradient elution using methanol and 0.1% formic acid in water. The calibration curves of all five analytes showed good linearity (R2>0.991). Accuracy and precision were within the required limits. The developed method could serve as an effective method for QC of A. tibetica. The investigated compounds were determined simultaneously for the first time in A. tibetica or any other plant.
Sigma-Aldrich
Ethyl acetate, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Ethyl acetate, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethyl acetate, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethyl acetate, JIS 1000, ≥99.5%, suitable for residue analysis