MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Acceptability and characterization of extruded pinto, navy and black beans.
Acceptability and characterization of extruded pinto, navy and black beans.
Journal of the science of food and agriculture (2014-10-10)
Courtney W Simons, Clifford Hall, Mehmet Tulbek, Mihiri Mendis, Taylor Heck, Samuel Ogunyemi
ABSTRACT
Consumption of dry beans has been relatively flat over the last decade. Creating new bean products may increase the consumption of beans and allow more consumers to obtain the health benefits of beans. In this study, pinto, navy and black beans were milled and the resulting flours extruded into puffs. Unflavored extruded puffs were evaluated by untrained panelists using a hedonic scale for appearance, flavor, texture and overall acceptability. The compositions of raw flours and extrudates were characterized. Sensory results indicated that all beans met or exceeded the minimum requirement for acceptability. Overall acceptability of navy and pinto beans was not significantly different, while acceptability of black bean puffs was significantly lower. Total protein (198-217 g kg(-1)) in extrudates was significantly different among the three beans. Total starch ranged from 398 to 406 g kg(-1) and was not significantly different. Resistant starch, total extractable lipid and raffinose contents were significantly reduced by extrusion. Extrusion did not affect crude fiber and phytic acid contents. The minimal effects on protein and fiber contents, the significant reduction in raffinose content and the acceptability of the unflavored extruded puffs support using various bean flours as ingredients in extruded puffed products.
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, suitable for HPLC
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, suitable for residue analysis
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, suitable for residue analysis
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Hexane, suitable for residue analysis, JIS 5000