MilliporeSigma
  • Signaling Rho-kinase mediates inflammation and apoptosis in T cells and renal tubules in cisplatin nephrotoxicity.
Signaling Rho-kinase mediates inflammation and apoptosis in T cells and renal tubules in cisplatin nephrotoxicity.
American journal of physiology. Renal physiology (2015-02-05)
Yuji Nozaki, Koji Kinoshita, Shoichi Hino, Tomohiro Yano, Kaoru Niki, Yasuaki Hirooka, Kazuya Kishimoto, Masanori Funauchi, Itaru Matsumura
ABSTRACT
Nephrotoxicity is a frequent complication of cisplatin-induced chemotherapy, in which T cells are known to promote acute kidney injury (AKI). Apoptosis and necrosis of tubules and inflammatory events also contribute to the nephrotoxicity. A delineation of the mechanisms that underlie the inappropriate renal and tubular inflammation can thus provide important insights into potential therapies for cisplatin-induced AKI. Rho-kinases are known to act as molecular switches controlling several critical cellular functions, including cell migration, cytokine production, and apoptosis. Here, we show that the Rho-kinase inhibitor fasudil attenuated cisplatin nephrotoxicity, resulting in less histological damage, improved renal function, and the infiltration of fewer leukocytes into the kidney. Renal nuclear factor-κB activation and apoptosis were reduced, and the expressions of proinflammatory renal cytokine and chemokine mRNA were decreased. Urinary and renal kidney injury molecule-1 (Kim-1) expression was also reduced, a finding that is consistent with diminished kidney injury. In the current study, we also showed that fasudil could be protective of the impaired tubules. In vitro, fasudil reduced the apoptosis (annexin-V+PI cells) and cytokine production (tumor necrosis factor+ cells) in T cells and the apoptosis (annexin-V+PI cells) and tubular damage (Kim-1+ cells) in proximal tubular cells by flow cytometric analysis. As Rho-kinase plays an important role in promoting cisplatin nephrotoxicity, inhibiting Rho-kinase may be a therapeutic strategy for preventing cisplatin-induced AKI.
Sigma-Aldrich
Brefeldin A, Ready Made Solution, from Penicillium brefeldianum, 10 mg/mL in DMSO
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, Molecular Biology
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid
Sigma-Aldrich
Propidium iodide solution