MilliporeSigma
  • Monitoring the formation of cholesterol oxidation products in model systems using response surface methodology.
Monitoring the formation of cholesterol oxidation products in model systems using response surface methodology.
Lipids in health and disease (2015-07-24)
Joong-Seok Min, Sang-Ok Lee, Muhammad Issa Khan, Dong Gyun Yim, Kuk-Hwan Seol, Mooha Lee, Cheorun Jo
ABSTRACT
Cholesterol oxidation products (COPs) are produced during thermal processing of animal origin foods and are considered to have negative health impacts. The model systems are helpful to understand the impact of various factors on oxidation changes in foods during cooking process. The study presented herein investigates the effects of pH, presence of unsaturated fatty acids, and heat on the formation of cholesterol oxidation products (COPs). Two model systems were designed to investigate the formation of cholesterol oxidation products in different lipid environments. The cholesterol oxides produced were quantified using gas chromatography. The level of cholesterol oxidation products decreased significantly at higher pH (above 5.8) and shorter heating time (3 h). The presence of unsaturated fatty acids (linoleic and oleic acids) significantly increased the amount of COPs under low-temperature heating conditions (100 °C and 1 h) but did not affect the production of COPs at higher temperature (150 °C). Increasing the temperature to 200 °C significantly increased the amount of COPs during the first hour of heating and this amount decreased upon further heating. The most frequently observed COPs in samples were α-epoxide, 20α-hydroxycholesterol, and 25-hydroxycholesterol. In conclusion, pH below 5.8, presence of unsaturated fatty acid, and high cooking temperature (>150 °C) leads to increased production of cholesterol oxidation products.
Product Description
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Acetone, suitable for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sigma-Aldrich
Acetone, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Acetone, suitable for chromatography, ≥99.8%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Silicic acid, BioReagent, suitable for column chromatography, 200-400 mesh (38 - 75 μm)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)