MilliporeSigma
  • Identification of the Protective Role of DJ-1 in Hypoglycemic Astrocyte Injury Using Proteomics.
Identification of the Protective Role of DJ-1 in Hypoglycemic Astrocyte Injury Using Proteomics.
Journal of proteome research (2015-06-10)
Jiangshan Deng, Fei Zhao, Xiaoyan Yu, Dawei Li, Yuwu Zhao
ABSTRACT
As a common complication of glycemic control in patients with diabetes, hypoglycemia often leads to brain dysfunction or damage. To identify new mechanisms underlying hypoglycemic brain injury, we determined the difference of protein expression profiles in brains between hypoglycemic rats and sham hypoglycemic controls by isobaric tags for relative and absolute quantitation (iTRAQ) analysis. Among the 89 deregulated proteins, DJ-1 protein (Park7) was verified to be upregulated following hypoglycemia insult in vivo and glucose deprivation in an astrocyte cell line (CTX-TNA2) cultured in vitro. Further studies indicated the pro-survival role of autophagy activation and impaired autophagy flux in CTX-TNA2 cells short of glucose. DJ-1 knockdown hindered the initiation of the autophagy process via the AMPK/mTOR pathway and aggravated cell death induced by glucose deficiency. Taken together, our results show that responsive overexpression of DJ-1 plays a protective role against hypoglycemic astrocyte injury partly mediated by the regulation of autophagy.
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Sucrose, SAJ first grade
Sigma-Aldrich
Sucrose, JIS special grade
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)