MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Does Glp-2 have a protective effect on cerebral ischemia/reperfusion model?
Does Glp-2 have a protective effect on cerebral ischemia/reperfusion model?
Turkish journal of medical sciences (2015-08-19)
Naci Topaloğlu, Gülsün Memi, Tuncay Kaner, Mustafa Deniz, Önder Şahin, Mustafa Güven, Murat Çoşar
ABSTRACT
To investigate the neuroprotective effects of glucagon-like peptide 2 (Glp-2), which increases cerebral blood flow, on the hippocampal complex after cerebral ischemia/reperfusion (I/R) injury in rats. Animals were randomized into 4 groups: sham, I/R + 0.9% NaCl, I/R + pre-Glp-2, and I/R + post-Glp-2. Cerebral ischemia was performed via the occlusion of the bilateral internal carotid artery for 40 min and continued with a reperfusion process. At the end of 6 h of reperfusion, animals were decapitated in all groups and brain tissues were removed. Malondialdehyde (MDA) and natural intracellular antioxidant glutathione (GSH) levels and myeloperoxidase (MPO) activities were measured in the left hippocampal tissue. The right hippocampal tissues of all group members were taken for histopathologic study. MDA levels and MPO activities increased from Group I to Group II and decreased from Group II to Groups III and IV. On the other hand, GSH levels were not significantly different among the groups. The number of apoptotic hippocampal tissue cells increased from Group I to Group II and decreased from Group II to Groups III and IV. Our preliminary study revealed that Glp-2 treatment may decrease oxidative damage from I/R in cerebral tissue.
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%