MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Influences of Ulva fasciata polysaccharide on the rheology and stabilization of cinnamaldehyde emulsions.
Influences of Ulva fasciata polysaccharide on the rheology and stabilization of cinnamaldehyde emulsions.
Carbohydrate polymers (2015-10-11)
Ping Shao, Jiamei Shao, Yike Jiang, Peilong Sun
ABSTRACT
Emulsifying properties of water soluble polysaccharides from Ulva fasciata (UFP) were evaluated in cinnamaldehyde/water emulsions in terms of droplet size distribution, rheological properties, visual phase separation, and zeta-potential. The cinnamaldehyde/water (10%, wt/wt) emulsions were formulated and stabilized by different concentrations of UFP (0.1-4%, wt/wt). The obtained emulsions showed monomodal droplet size distributions with average droplet size (D[3,2]) below 1.0μm, when 3% (wt/wt) UFP was added as the emulsifying agent under a homogenization pressure of 75MPa. The rheological properties and zeta-potential of the emulsions appeared to be dependent on the UFP concentration. Furthermore, the UFP exhibited better emulsifying and stabilizing properties in the investigated system when compared to other commercial polysaccharides of gum Arabic and gum Ghatti. The results also suggested that the emulsifying and stabilizing mechanism of the UFP may not only be ascribed to its surface-active protein moiety, but also to the hydrophobicity of the polysaccharide itself. These findings provided a theoretical basis for potential utilization of UFP as a novel hydrocolloid emulsifying agent.
Sigma-Aldrich
trans-Cinnamaldehyde, 97%
Sigma-Aldrich
trans-Cinnamaldehyde, ≥99%
Sigma-Aldrich
trans-Cinnamaldehyde, FCC, FG
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Cinnamaldehyde, natural, ≥95%, FG