MilliporeSigma
  • Raman spectroscopy for detecting supported planar lipid bilayers composed of ganglioside-GM1/sphingomyelin/cholesterol in the presence of amyloid-β.
Raman spectroscopy for detecting supported planar lipid bilayers composed of ganglioside-GM1/sphingomyelin/cholesterol in the presence of amyloid-β.
Physical chemistry chemical physics : PCCP (2015-08-11)
Zhiping Hu, Xiaoli Wang, Weirong Wang, Zhenlong Zhang, Huiping Gao, Yanli Mao
ABSTRACT
The aggregation and fibril formation of amyloid β(Aβ) peptides onto a ganglioside-GM1-containing lipid membrane is a cause of neurodegenerative diseases. The mechanism of the initial binding and the conformational changes of Aβ on the membrane should be clarified. Fluorescence microscopy and Raman spectroscopy have been performed to investigate the supporting planar lipid bilayers (SPBs) composed of ganglioside-GM1, sphingomyelin and cholesterol. It is demonstrated that the SPBs are in a liquid-crystalline state when placed on mica, and increasing the amount of ganglioside-GM1 can decrease the lateral interaction between the acyl chains of the SPBs. It has been found that Aβ(1-40) initially interacts with the galactose ring of the ganglioside-GM1 head group, leading to its binding and gradual aggregation on the membrane surface. The obvious change observed in Raman spectroscopy in the ν(C-H) region confirms that the hydrophobic C-terminal of Aβ(1-40) inserts itself into the hydrophobic part of the SPBs. The Raman data indicate that α-helix and β-sheet structures of Aβ(1-40) increase and coexist over longer time frames. Based on these results, a model was proposed to describe the mechanism of the conformational changes and the aggregation of Aβ(1-40) that are mediated by ganglioside-GM1-containing SPBs.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications