MilliporeSigma
  • Effects of human milk and formula on postprandial glycaemia and insulinaemia.
Effects of human milk and formula on postprandial glycaemia and insulinaemia.
European journal of clinical nutrition (2015-03-26)
C J Wright, F S Atkinson, N Ramalingam, A E Buyken, J C Brand-Miller
ABSTRACT
Consumption of formula in place of human milk may produce differences in postprandial glycaemia and insulinaemia that contribute to metabolic programming in the first year of life. The objective of the current study was to determine glycaemic and insulinaemic responses to human milk compared with a typical commercial formula, and then compare 11 other formulas. On separate mornings in random order, 10 healthy breastfeeding mothers consumed 25 g available carbohydrate portions of their own milk, a formula and reference food (25 g glucose on two occasions). In the second study, 10 different healthy subjects consumed 25 g available carbohydrate portions of 11 different commercial formulas and three reference foods (25 g glucose on three occasions). Fingerpick blood samples were taken at regular intervals over 2 h, and the glycaemic index (GI) and insulin index determined according to a standardised protocol. There were no significant differences in postprandial glycaemia or insulinaemia after human milk vs a typical formula (P = 0.3). Both produced a low GI (mean ± s.e.m.: 38 ± 7 vs 34 ± 7, respectively) and high insulin index (87 ± 14 vs 94 ± 16). The GI and insulin indices of the other formulas ranged from 18 ± 3 to 67 ± 6 and 53 ± 9 to 209 ± 33, respectively. Human milk and a typical formula elicit similar postprandial glycaemic and insulinaemic responses, but there is a wide range of responses to other formulas.
Sigma-Aldrich
β-Galactosidase from Aspergillus oryzae, ≥8.0 units/mg solid