MilliporeSigma
  • Reduced expression of SET7/9, a histone mono-methyltransferase, is associated with gastric cancer progression.
Reduced expression of SET7/9, a histone mono-methyltransferase, is associated with gastric cancer progression.
Oncotarget (2015-12-25)
Yoshimitsu Akiyama, Yuki Koda, Sun-Ju Byeon, Shu Shimada, Taketo Nishikawaji, Ayuna Sakamoto, Yingxuan Chen, Kazuyuki Kojima, Tatsuyuki Kawano, Yoshinobu Eishi, Dajun Deng, Woo Ho Kim, Wei-Guo Zhu, Yasuhito Yuasa, Shinji Tanaka
ABSTRACT
SET7/9, a histone methyltransferase, has two distinct functions for lysine methylation. SET7/9 methylates non-histone proteins, such as p53, and participates in their posttranslational modifications. Although SET7/9 transcriptionally activate the genes via H3K4 mono-methylation, its target genes are poorly understood. To clarify whether or not SET7/9 is related to carcinogenesis, we studied alterations of SET7/9 in gastric cancers (GCs). Among the 376 primary GCs, 129 cases (34.3%) showed loss or weak expression of SET7/9 protein compared to matched non-cancerous tissues by immunohistochemistry. Reduced SET7/9 expression was significantly correlated with clinical aggressiveness and worse prognosis. Knockdown of SET7/9 in GC cells markedly increased cell proliferation, migration and invasion. Expression of SREK1IP1, PGC and CCDC28B were inhibited in GC cells with SET7/9 knockdown, while matrix metalloproteinase genes (MMP1, MMP7 and MMP9) were activated. SET7/9 bound and mono-methylated H3K4 at the region of the approximately 4-6 kb upstream from the SREK1IP1 transcriptional start site and the promoters of PGC and CDC28B. Cell proliferation, migration and invasion, and expression of three MMPs were increased in GC cells with SREK1IP knockdown, which were similar to those of SET7/9 knockdown. These data suggest that SET7/9 has tumor suppressor functions, and loss of SET7/9 may contribute to gastric cancer progression.
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
MISSION® esiRNA, targeting human PGC
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®