MilliporeSigma
  • Antigen binding characteristics of immunoglobulin free light chains: crosslinking by antigen is essential to induce allergic inflammation.
Antigen binding characteristics of immunoglobulin free light chains: crosslinking by antigen is essential to induce allergic inflammation.
PloS one (2012-08-23)
Marco Thio, Tom Groot Kormelink, Marcel J Fischer, Bart R Blokhuis, Frans P Nijkamp, Frank A Redegeld
ABSTRACT
Beside the production of complete immunoglobulins IgG, IgE, IgA, IgM and IgD, consisting of tetrameric heterodimers of immunoglobulin heavy and light chains, B cells also secrete immunoglobulin free light chains (Ig-fLC). Previous studies showed that Ig-fLCs are able to induce immediate hypersensitivity reactions. It is apparent that recognition and binding of antigen are crucial steps in the onset of these inflammatory responses. In this study, the binding characteristics of Ig-fLC to antigen were further investigated using various biochemical approaches. In addition, we investigated whether antigen-mediated crosslinking of Ig-fLC is required to initiate allergic skin inflammation in vivo. Our study shows that binding of Ig-fLCs to antigen can be measured with different experimental setups. Surface plasmon resonance analysis showed real-time antigen binding characteristics. Specific antigen binding by Ig-fLCs was further detected using immunoblotting and ELISA. Using the ELISA-based assay, a binding affinity of 76.9±3.8 nM was determined for TNP-specific Ig-fLC. Antigen-induced ear swelling in mice passively sensitized with trinitrophenol-specific Ig-fLC was inhibited when multivalent antigen was combined with excess of monovalent antigen during challenge. We conclude that Ig-fLCs are able to interact with antigen, a prerequisite for antigen-specific cellular activation. In analogy to antigen-specific Fc receptor-induced mast cell activation, crosslinking of Ig-fLCs is necessary to initiate a local allergic response.
Sigma-Aldrich
TNP, ≥95% (HPLC)