MilliporeSigma
  • Asymmetric Synthesis of Substituted Thiolanes through Domino Thia-Michael-Henry Dynamic Covalent Systemic Resolution using Lipase Catalysis.
Asymmetric Synthesis of Substituted Thiolanes through Domino Thia-Michael-Henry Dynamic Covalent Systemic Resolution using Lipase Catalysis.
Advanced synthesis & catalysis (2014-03-24)
Yan Zhang, Pornrapee Vongvilai, Morakot Sakulsombat, Andreas Fischer, Olof Ramström
ABSTRACT
Dynamic systems based on consecutive thia-Michael and Henry reactions were generated and transformed using lipase-catalyzed asymmetric transformation. Substituted thiolane structures with three contiguous stereocenters were resolved in the process in high yields and high enantiomeric excesses.
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger