MilliporeSigma
  • FOXA1 acts upstream of GATA2 and AR in hormonal regulation of gene expression.
FOXA1 acts upstream of GATA2 and AR in hormonal regulation of gene expression.
Oncogene (2016-01-12)
J C Zhao, K-W Fong, H-J Jin, Y A Yang, J Kim, J Yu
ABSTRACT
Hormonal regulation of gene expression by androgen receptor (AR) is tightly controlled by many transcriptional cofactors, including pioneer factors FOXA1 and GATA2, which, however, exhibit distinct expression patterns and functional roles in prostate cancer. Here, we examined how FOXA1, GATA2 and AR crosstalk and regulate hormone-dependent gene expression in prostate cancer cells. Chromatin immunoprecipitation sequencing analysis revealed that FOXA1 reprograms both AR and GATA2 cistrome by preferably recruiting them to FKHD-containing genomic sites. By contrast, GATA2 is unable to shift AR or FOXA1 to GATA motifs. Rather, GATA2 co-occupancy enhances AR and FOXA1 binding to nearby ARE and FKHD sites, respectively. Similarly, AR increases, but not reprograms, GATA2 and FOXA1 cistromes. Concordantly, GATA2 and AR strongly enhance the transcriptional program of each other, whereas FOXA1 regulates GATA2- and AR-mediated gene expression in a context-dependent manner due to its reprogramming effects. Taken together, our data delineated for the first time the distinct mechanisms by which GATA2 and FOXA1 regulate AR cistrome and suggest that FOXA1 acts upstream of GATA2 and AR in determining hormone-dependent gene expression in prostate cancer.
Sigma-Aldrich
Anti-Androgen Receptor Antibody, from rabbit, purified by affinity chromatography