MilliporeSigma
  • Characterization of Water and a Model Lipophilic Compound in Human Stratum Corneum by NMR Spectroscopy and Equilibrium Sorption.
Characterization of Water and a Model Lipophilic Compound in Human Stratum Corneum by NMR Spectroscopy and Equilibrium Sorption.
Journal of pharmaceutical sciences (2016-09-28)
Chandana Kodiweera, Wayne J Romonchuk, Yuan Yang, Annette L Bunge
ABSTRACT
The stratum corneum (SC) is the outermost skin layer in humans and other mammals and the primary barrier to water loss and environmental exposure to chemicals and microorganisms. It consists of flattened, keratin-filled corneocytes surrounded by well-organized lipid layers. Human SC at varying degrees of hydration with and without addition of a model lipophilic compound, 2-(trifluoromethyl) benzonitrile (TFMB), was studied using proton (1H) and fluorine (19F) nuclear magnetic resonance techniques. Proton spectral analyses revealed that water mainly occupies the corneocytes in agreement with prior studies. Observations from 19F spectral and spin-lattice relaxation time (T1) analyses showed that TFMB is primarily present in the lipids with small amounts in water, which is located within the corneocytes. This is consistent with TFMB sorption, which was measured in SC with and without lipid extraction. The presence of TFMB within the corneocytes supports the hypothesis that transcellular diffusion of a lipophilic compound like TFMB may contribute to SC permeation.
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Sigma-Aldrich
N-p-Tosyl-L-phenylalanine chloromethyl ketone, ≥97% (TLC), powder
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Nα-Benzoyl-L-arginine ethyl ester hydrochloride, trypsin substrate