MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Frank-ter Haar syndrome protein Tks4 regulates epidermal growth factor-dependent cell migration.
Frank-ter Haar syndrome protein Tks4 regulates epidermal growth factor-dependent cell migration.
The Journal of biological chemistry (2012-07-26)
Gábor Bögel, Annamária Gujdár, Miklós Geiszt, Árpád Lányi, Anna Fekete, Szabolcs Sipeki, Julian Downward, László Buday
ABSTRACT
Mutations in the SH3PXD2B gene coding for the Tks4 protein are responsible for the autosomal recessive Frank-ter Haar syndrome. Tks4, a substrate of Src tyrosine kinase, is implicated in the regulation of podosome formation. Here, we report a novel role for Tks4 in the EGF signaling pathway. In EGF-treated cells, Tks4 is tyrosine-phosphorylated and associated with the activated EGF receptor. This association is not direct but requires the presence of Src tyrosine kinase. In addition, treatment of cells with LY294002, an inhibitor of PI 3-kinase, or mutations of the PX domain reduces tyrosine phosphorylation and membrane translocation of Tks4. Furthermore, a PX domain mutant (R43W) Tks4 carrying a reported point mutation in a Frank-ter Haar syndrome patient showed aberrant intracellular expression and reduced phosphoinositide binding. Finally, silencing of Tks4 was shown to markedly inhibit HeLa cell migration in a Boyden chamber assay in response to EGF or serum. Our results therefore reveal a new function for Tks4 in the regulation of growth factor-dependent cell migration.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
QCM Chemotaxis Cell Migration Assay, 24-well (8 µm), colorimetric, The QCM 24-well Migration Assay is ideal for the study of chemotaxis cell migration. The assay uses a 24-well plate with an 8 micron pore size, with colorimetric detection.
Millipore
Anti-V5 Agarose Affinity Gel antibody produced in mouse, purified immunoglobulin, clone V5-10
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse