MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Bis-pyridylethenyl benzene as novel backbone for amyloid-β binding compounds.
Bis-pyridylethenyl benzene as novel backbone for amyloid-β binding compounds.
Bioorganic & medicinal chemistry (2016-11-14)
Rob J A Nabuurs, Varsha V Kapoerchan, Athanasios Metaxas, Sarah Hafith, Maaike de Backer, Mick M Welling, Wim Jiskoot, Adrianus M C H van den Nieuwendijk, Albert D Windhorst, Herman S Overkleeft, Mark A van Buchem, Mark Overhand, Louise van der Weerd
ABSTRACT
Detection of cerebral β-amyloid (Aβ) by targeted contrast agents is of great interest for in vivo diagnosis of Alzheimer's disease (AD). Partly because of their planar structure several bis-styrylbenzenes have been previously reported as potential Aβ imaging agents. However, these compounds are relatively hydrophobic, which likely limits their in vivo potential. Based on their structures, we hypothesized that less hydrophobic bis-pyridylethenylbenzenes may also label amyloid. We synthesized several bis-pyridylethenylbenzenes and tested whether these compounds indeed display improved solubility and lower LogP values, and studied their fluorescent properties and Aβ binding characteristics. Bis-pyridylethenylbenzenes showed a clear affinity for Aβ plaques on both human and murine AD brain sections. Competitive binding experiments suggested a different binding site than Chrysamine G, a well-known stain for amyloid. With a LogP value between 3 and 5, most bis-pyridylethenylbenzenes were able to enter the brain and label murine amyloid in vivo with the bis(4-pyridylethenyl)benzenes showing the most favorable characteristics. In conclusion, the presented results suggest that bis-pyridylethenylbenzene may serve as a novel backbone for amyloid imaging agents.
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
2-Pyridinecarboxaldehyde, 99%
Sigma-Aldrich
3-Pyridinecarboxaldehyde, 98%