MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Efficient and rapid generation of large genomic variants in rats and mice using CRISMERE.
Efficient and rapid generation of large genomic variants in rats and mice using CRISMERE.
Scientific reports (2017-03-08)
Marie-Christine Birling, Laurence Schaeffer, Philippe André, Loic Lindner, Damien Maréchal, Abdel Ayadi, Tania Sorg, Guillaume Pavlovic, Yann Hérault
ABSTRACT
Modelling Down syndrome (DS) in mouse has been crucial for the understanding of the disease and the evaluation of therapeutic targets. Nevertheless, the modelling so far has been limited to the mouse and, even in this model, generating duplication of genomic regions has been labour intensive and time consuming. We developed the CRISpr MEdiated REarrangement (CRISMERE) strategy, which takes advantage of the CRISPR/Cas9 system, to generate most of the desired rearrangements from a single experiment at much lower expenses and in less than 9 months. Deletions, duplications, and inversions of genomic regions as large as 24.4 Mb in rat and mouse founders were observed and germ line transmission was confirmed for fragment as large as 3.6 Mb. Interestingly we have been able to recover duplicated regions from founders in which we only detected deletions. CRISMERE is even more powerful than anticipated it allows the scientific community to manipulate the rodent and probably other genomes in a fast and efficient manner which was not possible before.
Sigma-Aldrich
M2 medium, With HEPES, without penicillin and streptomycin, liquid, sterile-filtered, suitable for mouse embryo cell culture
Sigma-Aldrich
M16 Medium, With sodium bicarbonate and lactic acid, without penicillin and streptomycin, liquid, sterile-filtered