MilliporeSigma
  • Synergistic induction of apoptosis by salinomycin and gefitinib through lysosomal and mitochondrial dependent pathway overcomes gefitinib resistance in colorectal cancer.
Synergistic induction of apoptosis by salinomycin and gefitinib through lysosomal and mitochondrial dependent pathway overcomes gefitinib resistance in colorectal cancer.
Oncotarget (2015-10-16)
Zheng-Zhi Zou, Pei-Pei Nie, Ya-Wei Li, Ben-Xin Hou, Rui-Li, Xin-Peng Shi, Zhao-Kui Ma, Bao-Wei Han, Xiao-Yong Luo
ABSTRACT
Here, we showed the antibiotic salinomycin (SAL) combined with GEF exerted synergistic cytotoxicity effects in colorectal cancer cells irrespective of their EGFR and KRAS status, with a relatively low toxicity to normal cells. Additionally, combination of the two drugs overcame Ras-induced resistance and the acquired resistance to GEF. Further, we identified a new potential mechanism of this cooperative interaction by showing that GEF and SAL acted together to enhance production of reactive oxygen species (ROS), loss of mitochondrial membrane potential (MMP) and lysosomal membrane potential (LMP). And the ROS contributed the loss of MMP and LMP. We also found that GEF and SAL acted in concert to induce apoptosis via a mitochondrial-lysosomal cross-talk and caspase-independent pathway triggered by cathepsin B and D. Lastly, SAL in combination with GEF sensitized GEF-resistant cells to GEF in a nude mouse xenograft model. This novel combination treatment might provide a potential clinical application to overcome GEF resistance in colorectal cancer.
Sigma-Aldrich
Necrostatin-1, ≥98% (HPLC)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Necrostatin-5, ≥98% (HPLC), solid
Sigma-Aldrich
JC-1, powder or solid (Crystals)