MilliporeSigma
  • Alkyne activation and polyhedral reorganization in benzothiazolate-capped osmium clusters on reaction with diethyl acetylenedicarboxylate (DEAD) and ethyl propiolate.
Alkyne activation and polyhedral reorganization in benzothiazolate-capped osmium clusters on reaction with diethyl acetylenedicarboxylate (DEAD) and ethyl propiolate.
Dalton transactions (Cambridge, England : 2003) (2017-09-28)
Kh Mahid Uddin, Md Arshad H Chowdhury, Md Kamal Hossain, Shishir Ghosh, Derek A Tocher, Michael G Richmond, Shariff E Kabir
ABSTRACT
The reactivity of the face-capped benzothiazolate clusters HOs
Sigma-Aldrich
Ethyl propiolate, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Diethyl acetylenedicarboxylate, 95%
Pricing and availability is not currently available.