MilliporeSigma
  • GAS5, a non-protein-coding RNA, controls apoptosis and is downregulated in breast cancer.
GAS5, a non-protein-coding RNA, controls apoptosis and is downregulated in breast cancer.
Oncogene (2008-10-07)
M Mourtada-Maarabouni, M R Pickard, V L Hedge, F Farzaneh, G T Williams
ABSTRACT
Effective control of both cell survival and cell proliferation is critical to the prevention of oncogenesis and to successful cancer therapy. Using functional expression cloning, we have identified GAS5 (growth arrest-specific transcript 5) as critical to the control of mammalian apoptosis and cell population growth. GAS5 transcripts are subject to complex post-transcriptional processing and some, but not all, GAS5 transcripts sensitize mammalian cells to apoptosis inducers. We have found that, in some cell lines, GAS5 expression induces growth arrest and apoptosis independently of other stimuli. GAS5 transcript levels were significantly reduced in breast cancer samples relative to adjacent unaffected normal breast epithelial tissues. The GAS5 gene has no significant protein-coding potential but expression encodes small nucleolar RNAs (snoRNAs) in its introns. Taken together with the recent demonstration of tumor suppressor characteristics in the related snoRNA U50, our observations suggest that such snoRNAs form a novel family of genes controlling oncogenesis and sensitivity to therapy in cancer.
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Caspase-8 Inhibitor II, The Caspase-8 Inhibitor II controls the biological activity of Caspase-8. This small molecule/inhibitor is primarily used for Cancer applications.