MilliporeSigma
  • Rutin, a Quercetin Glycoside, Restores Chemosensitivity in Human Breast Cancer Cells.
Rutin, a Quercetin Glycoside, Restores Chemosensitivity in Human Breast Cancer Cells.
Phytotherapy research : PTR (2017-07-29)
Marcello Iriti, Robert Kubina, Andrea Cochis, Rita Sorrentino, Elena M Varoni, Agata Kabała-Dzik, Barbara Azzimonti, Arkadiusz Dziedzic, Lia Rimondini, Robert D Wojtyczka
ABSTRACT
Several studies have documented the ability of flavonoids to sensitize cancer cells to chemotherapeutics and reverse multidrug resistance by inhibition of efflux pumps (adenosine triphosphate-binding cassette transporters), apoptosis activation, and cell cycle arrest. In this study, the flavonoid rutin (quercetin 3-O-β-d-rutinoside) was investigated as chemosensitizer towards two different human epithelial breast cancer cell lines: (i) MB-MDA-231, selected as representative for triple-negative breast cancer and (ii) MCF-7 used as a well-characterized model of HER2-negative breast cancer. To assess the cytocompatibility of rutin against non-cancer cells, primary human mammary fibroblasts were used as control and non-target cells. In MDA-MB-231 cells, 20 μM rutin enhanced cytotoxicity related to cyclophosphamide and methotrexate. Rutin significantly (p < 0.05) increased the anticancer activity of both chemotherapeutics, at 24-48-72 h, and decreased the activity of the adenosine triphosphate-binding cassette transporters, namely, P-glycoprotein (P-gp) and breast cancer resistance protein (BCRP). Flow cytometry analysis showed 20 μM and 50 μM rutin arrested cell cycle at G2/M and G0/G1 phases, respectively, significantly promoting cell apoptosis. Rutin, via non-selective inhibition of P-gp and BCRP pumps, efficiently reverses multidrug resistance and restores chemosensitivity to cyclophosphamide and cyclophosphamide of human chemoresistant, triple-negative breast cancer cells, successfully arresting cell cycle progression. Copyright © 2017 John Wiley & Sons, Ltd.
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Ko143 hydrate, ≥98% (HPLC)