MilliporeSigma
Bioactivity Potential of
Frontiers in pharmacology (2017-11-01)
Anna Marchelak, Aleksandra Owczarek, Magdalena Matczak, Adam Pawlak, Joanna Kolodziejczyk-Czepas, Pawel Nowak, Monika A Olszewska
ABSTRACT
Flower extracts of
Sigma-Aldrich
3-Nitro-L-tyrosine, crystalline