MilliporeSigma
  • Functionalized lignin biomaterials for enhancing optical properties and cellular interactions of dyes.
Functionalized lignin biomaterials for enhancing optical properties and cellular interactions of dyes.
Biomaterials science (2017-08-24)
Anthony N Cauley, James N Wilson
ABSTRACT
We report a library of functionalized lignins and demonstrate their utility as nanocontainers for organic dyes in biologically relevant applications. Kraft lignin was modified via S
Sigma-Aldrich
3-Bromopropionic acid, 97%
Sigma-Aldrich
1-Iodobutane, 99%, contains copper as stabilizer
Sigma-Aldrich
1-Iododecane, 98%
Sigma-Aldrich
Benzyl bromide, reagent grade, 98%