MilliporeSigma
  • Obtention and characterization of the recombinant simian Interleukin-15 in Escherichia coli for the preclinical assessment of an IL-15-based therapeutic vaccine.
Obtention and characterization of the recombinant simian Interleukin-15 in Escherichia coli for the preclinical assessment of an IL-15-based therapeutic vaccine.
Preparative biochemistry & biotechnology (2017-08-18)
Yunier Rodríguez-Álvarez, Klaudia Martínez-Cordovez, Alexey Llopiz-Arzuaga, Yassel Ramos-Gómez, Vladimir Besada-Pérez, Dayana García-Lines, Alicia Santos-Savio
ABSTRACT
Recombinant simian IL-15 (siIL-15) was obtained for the preclinical assessment of an anti-human IL-15 vaccine. For this purpose, the cDNA from peripheral blood mononuclear cells of a Macaca fascicularis monkey was cloned into a pIL-2 vector. The siIL-15 was expressed in Escherichia coli strain W3110 as an insoluble protein which accounted for 13% of the total cellular proteins. Inclusion bodies were solubilized in an 8 M urea solution, which was purified by ion exchange and reverse phase chromatography up to 92% purity. The protein identity was validated by electrospray ionization-mass spectrometry, confirming the presence of the amino acids which distinguish the siIL-15 from human IL-15. The purified siIL-15 stimulates the proliferation of cytotoxic T-lymphocytes line (CTLL)-2 and Kit 225 cells with EC
Sigma-Aldrich
Triton X-100, laboratory grade