MilliporeSigma
  • Dietary B Vitamin Intake Is Associated with Lower Urinary Monomethyl Arsenic and Oxidative Stress Marker 15-F
Dietary B Vitamin Intake Is Associated with Lower Urinary Monomethyl Arsenic and Oxidative Stress Marker 15-F
The Journal of nutrition (2017-10-27)
Caitlin G Howe, Zhigang Li, Michael S Zens, Thomas Palys, Yu Chen, Jacqueline Y Channon, Margaret R Karagas, Shohreh F Farzan
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride