MilliporeSigma
Improvement of porcine islet culture with porcine serum.
Transplantation proceedings (1998-04-09)
K Sakamoto, E Hatakeyama, T Kenmochi, K Yamada, C Iwashita, T Asano, H Kashiwabara, T Yokoyama
Sigma-Aldrich
Porcine Serum, USA origin, sterile-filtered, suitable for cell culture