MilliporeSigma
  • Nonlinearity in protein assays by the Coomassie blue dye-binding method.
Nonlinearity in protein assays by the Coomassie blue dye-binding method.
Analytical biochemistry (1989-05-15)
A G Splittgerber, J Sohl
ABSTRACT
The Coomassie brilliant blue (CBB) method for protein determination takes advantage of the fact that a low-pH red form of CBB reverts to a blue form when CBB binds to protein. The increase in absorbance at 595 nm of a protein-dye mixture compared to a blank containing only the dye reagent has been used to estimate the total amount of protein present in the sample mixture. A disadvantage of this method of protein determination is that the assay plot of absorbance at 595 nm versus total protein is not linear.
Product Description
Supelco
Bradford Reagent, for 0.1-1.4 mg/ml protein