卫生监测
从原材料、供水到包装,食品生产环境很容易受到微生物污染。卫生监测与质量控制相结合,可确保食品和饮料生产设施的卫生条件,并将适用环境中产品和人员的交叉污染风险降至最低。
为了快速监测表面卫生和清洁效果,我们采用了众所周知的公认检测方法,如 ATP(三磷酸腺苷)、NAD/NADH(烟酰胺腺嘌呤二核苷酸)和蛋白质检测。这些方法都是可靠的指标。在微生物快速检测方面,我们的产品包括空气监测(主动和被动)、食源性病原体(PCR、免疫测定和 ELISA),以及 eMcollect Cloth、Swab &; Sponges。
在传统监测方面,我们使用各种选择性介质(浸渍拭子测试仪和接触式载玻片)来监测表面或液体样本中的卫生状况。此外,我们的一系列分析试剂和光度测量系统可通过对氯和季铵盐化合物进行半定量和定量检测来实现消毒控制。
产品
相关资源
- Brochure: eMCollect Sampling Solutions
Enhance your microbiological sampling capabilities with eMCollect products, featuring innovative designs that facilitate easy use and accurate monitoring of cleaning effectiveness.
- Food & Beverage Safety Brochure: Microbial QC Solutions
Discover efficient workflows and reliable testing solutions designed to enhance quality control in the food and beverage industry.
- Application Note: New LED Verification Pen
Learn about the advantages of the new LED Pen, featuring dual control measurement & battery status indicators for enhanced operational efficiency.
- Flyer: eMcollect Sterile Sampling Device
eMcollect swab cloths and sponges comply with ISO 17604 and ISO 18593 standards and are effective in collecting environmental monitoring samples. Download the flyer to know more.
- Flyer: FLASH® Allergen Indicator Protein (AIP) Test for “Big 9”
FLASH® AIP Test, a rapid and reliable protein indicator test that can detect allergenic proteins from “Big 9” including sesame
- Application Note: FLASH® RT Allergen Indicator Total Protein Test
FLASH® RT is designed for use at room temperature and can detect as little as 20 µg of total protein within 10 minutes.
- Application Note: ATP Testing Comparison on Disinfectants
The impact of disinfectants & sanitizers on the ATP signal is studied using a panel of industrial products commonly applied in the food, beverage and other regulated industries.
- Application Note: FREE vs Total ATP
Total ATP is the sum of free ATP and intracellular ATP. ATP is permanently renewed and recycled within the cell, but its production immediately stops when the cell dies.
- Brochure: MVP ICON® ATP Testing System
MVP ICON® sampling devices are designed to compensate for variations in swabbing technique, so no matter how many technicians you have or the frequency of staff turnover, you can be assured of accurate, consistent readings.
- Application Note: HY-LiTE® Testing of Paints and Coatings Samples
Regular microbial monitoring of paints and coatings is important to ensure timely action that affects quality and processes.
- Application Note: HY-LiTE® Surface Testing for Biofilm Contamination
The presence of biofilm in a system may sometimes be diagnosed by elevated readings of extra-cellular (Free) ATP and/or other metabolites in the liquid.
- Application Note: HY-LiTE® Plus – Testing of Biocide Efficacy
This document describes the method to demonstrate the speed and mode of action of different Biocides in water samples naturally contaminated or spiked with bacterial cultures.
- Application Note: Suspended Reading of HY-LiTE® Tests
The HY-LiTE® Liquid Test Pens are ready-to-use pens that measure total ATP in liquid samples using the HY-LiTE® 2/2A luminometer.
- Application Note: Suitability of MC Media Pad® for Hygiene Monitoring
The study focused on usage of MC‑Media Pad® in passive air, personnel and surface monitoring in comparison to standard test methods.
- Brochure: Swab & Sampler Test Kits for Quality Control
This technique eliminates the cumbersome and expensive hardware and materials associated with traditional testing techniques.
- Flyer: Envirocheck® Dip Slides
Envirocheck® Dip Slides provide semi-quantitative detection of microorganisms on surfaces and in liquids.
- Article: Navigating Disinfection Control in Food and Beverage Manufacturing
了解消毒控制以及如何在众多选择中挑选合适的消毒剂,不同的选择基于光度法和反射法的方法和仪器。
使用 eMcollect 提高卫生标准
eMcollect 提供一系列无菌设备,专门用于对食品行业的工作表面进行有效采样。
eMcollect无菌采样设备的主要特点使其成为环境监测最通用的解决方案:
- 布拭子尺寸为34 x 16厘米,具有蜂窝状结构,可有效采样。
- 布拭子和海绵有多种选择,包括带中和剂的布拭子和海绵,用于监测清洁效果。
- 不同配置的海绵,包括用于难以触及区域的带棒海绵。
快速方法
| Product | Target(s) | How to Measure | Instrumentation required? | 读取结果的时间(TTR) | 检测储存和; 保存期 |
|---|---|---|---|---|---|
| MVP ICON® System | ATP、电导率和浓度、pH 值、温度 | ATP 拭子在发光仪中读取;其他参数通过可选探针读取 | 18 秒 | 室温 | |
| HY-LiTE® 2 系统
| 表面、液体、工业样品和 A1 喷气燃料中的 ATP | 在发光仪中读取特定应用的 ATP 笔 | 是 | 16秒 | 2-8°C |
| <FLASH® RT 比色测定 | 包括过敏原在内的总蛋白 | 蛋白拭子比色检测(有/无) | 无 | 10±4分钟 | 室温 |
| HY-RiSE® 比色测定 | NAD/NADH及其磷酸盐 | 否 | 4 - 5 分钟 | 室温或 2-8 °C |
传统方法
| 产品 | 目标 | 如何测量 | 所需仪器? | 读取结果所需时间(TTR) | 检测储存和保质期 |
|---|---|---|---|---|---|
| 浸渍和拭子测试仪拭子测试仪拭子测试仪 | 菌落总数、大肠菌群、酵母菌和霉菌 | 培养后单面微生物菌落计数 | 否,培养箱除外 | 24-72小时 | 室温 |
| Envirocheck® 浸渍载玻片 | 总计数、肠杆菌、 大肠杆菌、大肠菌群 | ||||
| 培养后 A 面和 B 面不同微生物菌落的计数 | 没有。td>否,培养箱除外 | 24 小时至 7 天 | < 15 °C |
相关视频
如要继续阅读,请登录或创建帐户。
暂无帐户?为方便客户阅读,本页通过机器进行翻译。我们已尽力确保机器翻译的准确性,但机器翻译并非尽善尽美。如果您对机器翻译内容不满意,请参阅英文版本。