Skip to Content
MilliporeSigma

Skip To

ABC336

Anti-MORF4 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

MORF4 Antibody, MORF4, Mortality factor 4, Cellular senescence-related protein 1, SEN1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
$477.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IHC, WB
Citations:
-

$477.00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunofluorescence: suitable, immunohistochemistry: suitable, western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MORF4(10934)

General description

Mortality factor 4 (MORF4) is a component of the NuA4 histone acetyltransferase (HAT) complex which is involved in transcriptional activation of select genes typically by acetylation of nucleosomal histones H4 and H2A. NuA4 complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, DNA repair and apoptosis. MORF4 is localized to the nucleus and has transcription factor-like motifs such as helix-loop-helix and a leucine zipper motif. Additionally, MORF4 has been shown to bind to the transcription corepressors mSin3A and TLE, suggesting that together, these complexes may play a role in transcriptional repression of genes that lead to cellular senescence.
~33 kDa observed

Immunogen

KLH-conjugated linear peptide corresponding to human MORF4.

Application

Anti-MORF4 Antibody is a highly specific rabbit polyclonal antibody, that targets Mortality factor 4 & has been tested in western blotting, IHC & Immunofluorescence.
Immunohistochemistry Analysis: 5 µg/mL from a representative lot detected MORF4 in human brain tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected MORF4 in HepG2 cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
K562 cell lysate
Evaluated by Western Blotting in K562 cell lysate

Western Blotting Analysis: 2 µg/mL of this antibody detected MORF4 in 15 µg of K562 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABE387SAB2108065HPA027097
description

from rabbit, purified by affinity chromatography

description

serum, from rabbit

description

affinity isolated antibody

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

serum

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Gene Information

human ... MORF4(10934)

Gene Information

human ... ATF4(468)

Gene Information

human ... SURF4(6836)

Gene Information

human ... MTERFD2(130916)

technique(s)

immunohistochemistry: suitable, immunofluorescence: suitable, western blot: suitable

technique(s)

western blot: suitable, ChIP: suitable

technique(s)

immunoblotting: suitable

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunohistochemistry: 1:50-1:200, immunofluorescence: 0.25-2 μg/mL


Still not finding the right product?

Try our Product Selector Tool to narrow your options


WGK

WGK 2

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters