Skip to Content
MilliporeSigma
Corticosterone solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Corticosterone solution

Corticosterone solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C21H30O4
CAS Number:
Molecular Weight:
346.46

Products

Product Category
Brand
Manufacturer
Physical Form
Quality Segment
Application
Special Grade
Technique