MilliporeSigma
Role of xanthine oxidoreductase as an antimicrobial agent.
Infection and immunity (2004-08-24)
Hannah M Martin, John T Hancock, Vyv Salisbury, Roger Harrison
Sigma-Aldrich
Xanthine Oxidase from bovine milk, Grade IV, ammonium sulfate suspension, ≥0.1 units/mg protein