MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Mechanisms of antihyperglycaemic action of efaroxan in mice: time for reappraisal of α2A-adrenergic antagonism in the treatment of type 2 diabetes?
Mechanisms of antihyperglycaemic action of efaroxan in mice: time for reappraisal of α2A-adrenergic antagonism in the treatment of type 2 diabetes?
Diabetologia (2012-08-18)
Z Lehner, K Stadlbauer, I Adorjan, I Rustenbeck, M Belz, A Fenzl, V A M de Cillia, D Gruber, L Bauer, K Frobel, B Brunmair, A Luger, C Fürnsinn
ABSTRACT
Inspired by recent speculation about the potential utility of α(2A)-antagonism in the treatment of type 2 diabetes, the study examined the contribution of α(2)-antagonism vs other mechanisms to the antihyperglycaemic activity of the imidazoline (±)-efaroxan. Effects of the racemate and its pure enantiomers on isolated pancreatic islets and beta cells in vitro, as well as on hyperglycaemia in vivo, were investigated in a comparative manner in mice. In isolated perifused islets, the two enantiomers of efaroxan were equally potent in counteracting inhibition of insulin release by the ATP-dependent K(+) (K(ATP)) channel-opener diazoxide but (+)-efaroxan, the presumptive carrier of α(2)-antagonistic activity, was by far superior in counteracting inhibition of insulin release by the α(2)-agonist UK14,304. In vivo, (+)-efaroxan improved oral glucose tolerance at 100-fold lower doses than (-)-efaroxan and, in parallel with observations made in vitro, was more effective in counteracting UK14,304-induced than diazoxide-induced hyperglycaemia. The antihyperglycaemic activity of much higher doses of (-)-efaroxan was associated with an opposing pattern (i.e. with stronger counteraction of diazoxide-induced than UK14,304-induced hyperglycaemia), which implicates a different mechanism of action. The antihyperglycaemic potency of (±)-efaroxan in mice is almost entirely due to α(2)-antagonism, but high doses can also lower blood glucose via another mechanism. Our findings call for reappraisal of the possible clinical utility of α(2A)-antagonistic compounds in recently identified subpopulations of patients in which a congenitally higher level of α(2A)-adrenergic activation contributes to the development and pathophysiology of type 2 diabetes.
Product Description
Diazoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Efaroxan hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Diazoxide
Sigma-Aldrich
UK 14,304