MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • By inhibiting Src, verapamil and dasatinib overcome multidrug resistance via increased expression of Bim and decreased expressions of MDR1 and survivin in human multidrug-resistant myeloma cells.
By inhibiting Src, verapamil and dasatinib overcome multidrug resistance via increased expression of Bim and decreased expressions of MDR1 and survivin in human multidrug-resistant myeloma cells.
Leukemia research (2013-11-19)
Masanobu Tsubaki, Makiko Komai, Tatsuki Itoh, Motohiro Imano, Kotaro Sakamoto, Hirotaka Shimaoka, Tomoya Takeda, Naoki Ogawa, Kenji Mashimo, Daiichiro Fujiwara, Junji Mukai, Katsuhiko Sakaguchi, Takao Satou, Shozo Nishida
ABSTRACT
The calcium channel blocker verapamil inhibits the transport function of multidrug resistance protein 1 (MDR1). Although verapamil acts to reverse MDR in cancer cells, the underlying mechanism remains unclear. In the present study, we investigated the mechanism of reversing MDR by verapamil in anti-cancer drug-resistant multiple myeloma (MM) cell lines. We found that verapamil suppresses MDR1 and survivin expressions and increases Bim expression via suppression of Src activation. Furthermore, dasatinib reversed the drug-resistance of the drug-resistant cell lines. These findings suggest that Src inhibitors are potentially useful as an anti-MDR agent for the treatment of malignant tumor cells.
Product Description
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Supelco
Verapamil hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder