MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Methylamine utilization via the N-methylglutamate pathway in Methylobacterium extorquens PA1 involves a novel flow of carbon through C1 assimilation and dissimilation pathways.
Methylamine utilization via the N-methylglutamate pathway in Methylobacterium extorquens PA1 involves a novel flow of carbon through C1 assimilation and dissimilation pathways.
Journal of bacteriology (2014-09-17)
Dipti D Nayak, Christopher J Marx
ABSTRACT
Methylotrophs grow on reduced single-carbon compounds like methylamine as the sole source of carbon and energy. In Methylobacterium extorquens AM1, the best-studied aerobic methylotroph, a periplasmic methylamine dehydrogenase that catalyzes the primary oxidation of methylamine to formaldehyde has been examined in great detail. However, recent metagenomic data from natural ecosystems are revealing the abundance and importance of lesser-known routes, such as the N-methylglutamate pathway, for methylamine oxidation. In this study, we used M. extorquens PA1, a strain that is closely related to M. extorquens AM1 but is lacking methylamine dehydrogenase, to dissect the genetics and physiology of the ecologically relevant N-methylglutamate pathway for methylamine oxidation. Phenotypic analyses of mutants with null mutations in genes encoding enzymes of the N-methylglutamate pathway suggested that γ-glutamylmethylamide synthetase is essential for growth on methylamine as a carbon source but not as a nitrogen source. Furthermore, analysis of M. extorquens PA1 mutants with defects in methylotrophy-specific dissimilatory and assimilatory modules suggested that methylamine use via the N-methylglutamate pathway requires the tetrahydromethanopterin (H4MPT)-dependent formaldehyde oxidation pathway but not a complete tetrahydrofolate (H4F)-dependent formate assimilation pathway. Additionally, we present genetic evidence that formaldehyde-activating enzyme (FAE) homologs might be involved in methylotrophy. Null mutants of FAE and homologs revealed that FAE and FAE2 influence the growth rate and FAE3 influences the yield during the growth of M. extorquens PA1 on methylamine.
Sigma-Aldrich
Methylamine-15N hydrochloride, 98 atom % 15N
Carbon, foil, 25x25mm, thickness 0.5mm, pyrolytic graphite, 99.99%
Carbon, rod, 100mm, diameter 1.0mm, graphite, 99.95%
Carbon, rod, 50mm, diameter 0.5mm, graphite, 99.95%
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Carbon, foil, 50x50mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
Carbon, foil, 100x100mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 50mm, diameter 1.0mm, graphite, 99.95%
Carbon, rod, 150mm, diameter 5.0mm, graphite, 100%
Carbon, foil, 12.5x12.5mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 100mm, diameter 3.0mm, graphite, 100%
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Millipore
Activated charcoal, suitable for GC
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size