MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Dynamics and rheology of nonpolar bijels.
Soft matter (2015-06-09)
Lian Bai, John W Fruehwirth, Xiang Cheng, Christopher W Macosko
ABSTRACT
Bicontinuous, interfacially jammed, emulsion gels (bijels) are a novel class of materials composed of two immiscible phases with interpenetrating domains that are stabilized by a monolayer of colloidal particles at the interface. However, existing bijel systems so far all consist of at least one polar fluid, which is believed to be essential to induce electrostatic repulsion for stabilizing interfacial particles. It is not known whether two nonpolar fluids can form a bijel. Here, we experimentally achieve a bijel using styrene trimer and low molecular weight polybutene-two nonpolar fluids that are similar to polymer blends, which are important in technical applications. By combining laser scanning confocal microscopy, cryo-SEM and rheology measurement, we systematically investigate the dynamics and rheology of this nonpolar bijel. In contrast to previous studies on polar bijels, we observe the formation of localized regions of high particle concentration or "particle patches" on the interface which assemble during coarsening. We also provide the first quantitative relation between the morphology of a bijel, the interfacial particle coverage and the shear modulus during bijel coarsening. Moreover, we reveal a previously unnoticed increase in the elastic modulus of bijels that can be attributed to the rearrangement of interfacial particles at long time scales. In addition, we also found a hydrophobic particle framework that survives after the direct remixing of the nonpolar bijel. Our study provides important insights into the formation of bijels and is the first step to explore the missing link between polar bijels and particle-stabilized bicontinuous polymer blends.
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.99 atom % 12C
Sigma-Aldrich
Rhodamine B isothiocyanate, BioReagent, mixed isomers, suitable for protein labeling
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, foil, 50x50mm, thickness 4.0mm, glassy carbon