MilliporeSigma
  • Small-molecule ligands bind to a distinct pocket in Ras and inhibit SOS-mediated nucleotide exchange activity.
Small-molecule ligands bind to a distinct pocket in Ras and inhibit SOS-mediated nucleotide exchange activity.
Proceedings of the National Academy of Sciences of the United States of America (2012-03-21)
Till Maurer, Lindsay S Garrenton, Angela Oh, Keith Pitts, Daniel J Anderson, Nicholas J Skelton, Benjamin P Fauber, Borlan Pan, Shiva Malek, David Stokoe, Mary J C Ludlam, Krista K Bowman, Jiansheng Wu, Anthony M Giannetti, Melissa A Starovasnik, Ira Mellman, Peter K Jackson, Joachim Rudolph, Weiru Wang, Guowei Fang
ABSTRACT
The Ras gene is frequently mutated in cancer, and mutant Ras drives tumorigenesis. Although Ras is a central oncogene, small molecules that bind to Ras in a well-defined manner and exert inhibitory effects have not been uncovered to date. Through an NMR-based fragment screen, we identified a group of small molecules that all bind to a common site on Ras. High-resolution cocrystal structures delineated a unique ligand-binding pocket on the Ras protein that is adjacent to the switch I/II regions and can be expanded upon compound binding. Structure analysis predicts that compound-binding interferes with the Ras/SOS interactions. Indeed, selected compounds inhibit SOS-mediated nucleotide exchange and prevent Ras activation by blocking the formation of intermediates of the exchange reaction. The discovery of a small-molecule binding pocket on Ras with functional significance provides a new direction in the search of therapeutically effective inhibitors of the Ras oncoprotein.
Sigma-Aldrich
DCAI, ≥95% (HPLC)