MilliporeSigma
  • CeFra-seq reveals broad asymmetric mRNA and noncoding RNA distribution profiles in
CeFra-seq reveals broad asymmetric mRNA and noncoding RNA distribution profiles in
RNA (New York, N.Y.) (2017-10-29)
Louis Philip Benoit Bouvrette, Neal A L Cody, Julie Bergalet, Fabio Alexis Lefebvre, Cédric Diot, Xiaofeng Wang, Mathieu Blanchette, Eric Lécuyer
ABSTRACT
Cells are highly asymmetrical, a feature that relies on the sorting of molecular constituents, including proteins, lipids, and nucleic acids, to distinct subcellular locales. The localization of RNA molecules is an important layer of gene regulation required to modulate localized cellular activities, although its global prevalence remains unclear. We combine biochemical cell fractionation with RNA-sequencing (CeFra-seq) analysis to assess the prevalence and conservation of RNA asymmetric distribution on a transcriptome-wide scale in
Sigma-Aldrich
Shields and Sang M3 Insect Medium, With L-glutamine, without potassium bicarbonate, powder, suitable for insect cell culture
Sigma-Aldrich
Select Yeast Extract, powder, BioReagent, suitable for cell culture, suitable for insect cell culture