MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
MilliSentials™
Lab Labeling System
Streamline Sample, Reagent, & Chemical Labeling
Watch this video for an introduction on the capabilities of the MilliSentials™ Lab Labeling System.
Label it Right
Unknown samples in your -80 °C freezer? Trouble deciphering your labmate’s handwriting? Frustrated with labels that fade? The MilliSentials™ Lab Labeling System streamlines sample, reagent, and chemical identification with impervious, adaptable labels tailored for research labs.  
  • Printed, user-defined labels for greater legibility
  • Cryogenic and chemically-resistant labels designed for lab environments
  • Temperature resistance from -196 °C (liquid nitrogen) to 100 °C
  • Chemical resistance to water, ethanol, IPA, DMSO, methanol, acetonitrile, bleach, acetone, xylene, and toluene
  • Pre-defined templates for label creation
  • Scannable 1D and 2D barcode creation, as well as parent/child labels, help to further optimize your sample tracking process
MilliSentials™ Labels for slides, plates, flasks, tubes, and bottles
Anything Goes
The MilliSentials™ Lab Labeling System was designed specifically for the research lab, with adaptable labels that can be adjusted to fit any vessel or container.
  • Microscope slides
  • Flasks
  • Dishes
  • Plates
  • Beakers
  • Bottles
  • Vials
  • Tubes
  • More tubes
Request More Information
Thank you for your interest in our MilliSentials™ Lab Labeling System. Please complete the below form and we will be in touch.
Fields indicated by an * are required.
Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500