Skip to Content
MilliporeSigma

Skip To

AV49592

Anti-CACHD1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Cache domain containing 1, Anti-KIAA1573, Anti-RP4-655E10.1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
$617.00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
4

$617.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

137 kDa

species reactivity

mouse, horse, human, dog, guinea pig, rabbit, bovine, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CACHD1(57685)

General description

The gene CAHD1 (Cache domain-containing protein 1) is mapped to human chromosome 1p31.3.

Immunogen

Synthetic peptide directed towards the N terminal region of human CACHD1

Application

Anti-CACHD1 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

CACHD1 (Cache domain-containing protein 1) is a calcium channel protein that is important for excitation-contraction coupling. It is differentially regulated in humans suffering from parkinson disease. CACHD1 is up-regulated in human embryonic stem cells and is down-modulated upon differentiation.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: HKFRCKGSYEHRSRPIYVSTVRPQSKHIVVILDHGASVTDTQLQIAKDAA

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB5700260HPA022236HPA039046
description

affinity isolated antibody

description

-

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

— or —

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters