MilliporeSigma
Unmatched TFF Performance for mAbs, ADCs, and Viral Vectors
Single-use, High-yield Filtration Made Simple
Accelerate Your Bioprocessing with Pellicon® Capsules
Pellicon® Capsules are single-use tangential flow filtration (TFF) devices featuring Ultracel® membrane, optimized for efficient concentration and purification of biologics like antibodies, viral vectors, and ADCs while minimizing contamination risks. Designed for faster, smarter bioprocessing, they offer plug ‘n play, high-yield filtration with superior flux and scalability, enabling rapid setup and seamless process integration.
Experience the benefits of Pellicon® Capsules for yourself—click here to request your free sample.
Why Pellicon® Capsules?
  • Presterilized & Ready-to-Use – Setup in minutes, no cleaning required.
  • High Flux & Recovery – Faster processing and greater yields.
  • Scalable & Reliable – Consistent performance across all scales.
Tailored to Your Process
ADCs: Closed, single-use design enhances safety and efficiency.
Tangential flow filtration (TFF) workflows using Pellicon® Capsules enable efficient removal of solvents and other process impurities, while protecting operators from risks associated with exposure to the highly potent ADC intermediates. Learn more.
Viral Vectors: Up to 3× faster processing than hollow fibers.
This study compared performance of Pellicon® Capsules with hollow fiber modules for volume reduction and diafiltration of AAV post clarification. Results showed that although yield was equivalent between Pellicon® Capsules and hollow fiber modules, the Pellicon® Capsules achieved higher and more stable flux which resulted in faster processing and lower membrane area requirements, improving the efficiency, costs, and footprint of the operation.
mAbs: Higher final concentrations, no recirculation needed.
Single pass tangential flow filtration (SPTFF) enables intensified mAb downstream processing by efficiently concentrating process streams without recirculation. SPTFF eliminates the need for hold tanks and improves footprint without compromising yield. This article discusses SPTFF concentration with Pellicon® Capsules and highlights how it can be easily scaled from small- to large-scale operations.
Streamline your workflow and boost productivity. Try Pellicon® Capsules today!
Fill out the form below to request your free sample.*
VALID ONLY in the United States, Canada, Austria, Belgium, Bulgaria, Croatia, Republic of Cyprus, Czech Republic, Denmark, Estonia, Finland, France, Germany, Greece, Japan, Hungary, India, Ireland, Italy, Latvia, Lithuania, Luxembourg, Malta, Netherlands, Norway, Poland, Portugal, Romania, Singapore, Slovakia, Slovenia, South Korea, Spain, Sweden, Switzerland, Taiwan, Thailand, the UK and Vietnam. VOID WHERE PROHIBITED BY LAW.
Pellicon® Capsule Trial Offer Official Rules
*One sample of Pellicon® Capsule 0.1 m2 with 30 kDa Ultracel® Membrane and C screen (PCC030C01) OR Pellicon® Capsule 0.1 m2 with 30 kDa Ultracel® Membrane, C screen, and AseptiQuik® G Connectors (PCC030C01C) OR Pellicon® Capsule 0.1 m2 with 100 kDa Ultracel® Membrane and C screen (PCC100C01) OR Pellicon® Capsule 0.1 m2 with 100 kDa Ultracel® Membrane, C screen, and AseptiQuik® G Connectors (PCC100C01C) OR Pellicon® Capsule 0.1 m2 with 300 kDa Ultracel® Membrane and C screen (PCC300C01) OR Pellicon® Capsule 0.1 m2 with 300 kDa Ultracel® Membrane, C screen, and AseptiQuik® G Connectors (PCC300C01C)