Skip to Content
MilliporeSigma

Skip To

GF26399997

Nickel

rod, 8.0 mm diameter, length 100 mm, high purity 99.99+%

Synonym(s):

NI007958

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
$950.00

About This Item

Linear Formula:
Ni
CAS Number:
Molecular Weight:
58.69
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141729
MDL number:
eCl@ss:
38190701

$950.00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99.99%, ±99.99%

form

rod

manufacturer/tradename

Goodfellow 263-999-97

resistivity

6.97 μΩ-cm, 20°C

L

100 mm

diameter

8.0 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
577995GF04564470GF97876445
description

rod, 8.0 mm diameter, length 100 mm, high purity 99.99+%

description

nanopowder, <100 nm avg. part. size, ≥99% trace metals basis

description

foil, 50mm disks, thickness 0.01mm, 99.95%

description

wire reel, 500m, diameter 0.25mm, as drawn, 99%

form

rod

form

nanopowder

form

foil

form

wire

assay

≥99.99%, ±99.99%

assay

≥99% trace metals basis

assay

99.95%

assay

≥99%

manufacturer/tradename

Goodfellow 263-999-97

manufacturer/tradename

-

manufacturer/tradename

Goodfellow 045-644-70

manufacturer/tradename

Goodfellow 978-764-45

density

8.9 g/mL at 25 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

resistivity

6.97 μΩ-cm, 20°C

resistivity

6.97 μΩ-cm, 20°C

resistivity

6.97 μΩ-cm, 20°C

resistivity

6.97 μΩ-cm, 20°C

mp

1453 °C (lit.)

mp

1453 °C (lit.)

mp

1453 °C (lit.)

mp

1453 °C (lit.)


Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters