Skip to Content
MilliporeSigma

Skip To

GF29337352

Aluminum-magnesium alloy, Al97Mg3

500mm tube, 1.0mm outside diameter, 0.075mm wall thickness, 0.85mm inside diameter, as drawn

Synonym(s):

AL017200

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.23
UNSPSC Code:
12352311
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

tubes

composition

Al97Mg3

reaction suitability

core: aluminum, core: magnesium

manufacturer/tradename

Goodfellow 293-373-52

L × wall thickness

500 mm × 0.075 mm

O.D. × I.D.

1.0 mm × 0.85 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF88903558GF25034699GF18118391
description

500mm tube, 1.0mm outside diameter, 0.075mm wall thickness, 0.85mm inside diameter, as drawn

description

1000mm tube, 0.53mm outside diameter, 0.075mm wall thickness, 0.38mm inside diameter, as drawn

description

200mm tube, 1.0mm outside diameter, 0.075mm wall thickness, 0.85mm inside diameter, as drawn

description

200mm tube, 0.74mm outside diameter, 0.065mm wall thickness, 0.61mm inside diameter, as drawn

form

tubes

form

tubes

form

tubes

form

tubes

manufacturer/tradename

Goodfellow 293-373-52

manufacturer/tradename

Goodfellow 889-035-58

manufacturer/tradename

Goodfellow 250-346-99

manufacturer/tradename

Goodfellow 181-183-91

O.D. × I.D.

1.0 mm × 0.85 mm

O.D. × I.D.

-

O.D. × I.D.

1.0 mm × 0.85 mm

O.D. × I.D.

0.74 mm × 0.61 mm

L × wall thickness

500 mm × 0.075 mm

L × wall thickness

-

L × wall thickness

200 mm × 0.075 mm

L × wall thickness

-

reaction suitability

core: magnesium, core: aluminum

reaction suitability

core: aluminum, core: magnesium

reaction suitability

core: magnesium, core: aluminum

reaction suitability

core: aluminum, core: magnesium

composition

Al97Mg3

composition

Al97Mg3

composition

Al97Mg3

composition

Al97Mg3


Still not finding the right product?

Explore all of our products under Aluminum-magnesium alloy, Al97Mg3




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters