Skip to Content
MilliporeSigma

Skip To

GF57849014

Polyamide - Nylon 6

(PA 6), sheet, thickness 5 mm, size 300 × 300 mm

Synonym(s):

AM303050

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.23
UNSPSC Code:
12352100
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

sheet

manufacturer/tradename

Goodfellow 578-490-14

size

300 × 300 mm

thickness

5 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF86862796GF04550880GF00705495
description

(PA 6), sheet, thickness 5 mm, size 300 × 300 mm

description

(PA 6), sheet, thickness 5 mm, size 150 × 150 mm

description

(PA 6), sheet, thickness 10 mm, size 500 × 500 mm

description

(PA 6), sheet, thickness 3 mm, size 500 × 500 mm

manufacturer/tradename

Goodfellow 578-490-14

manufacturer/tradename

Goodfellow 868-627-96

manufacturer/tradename

Goodfellow 045-508-80

manufacturer/tradename

Goodfellow 007-054-95

form

sheet

form

sheet

form

sheet

form

sheet

thickness

5 mm

thickness

5 mm

thickness

10 mm

thickness

3 mm

size

300 × 300 mm

size

150 × 150 mm

size

500 × 500 mm

size

500 × 500 mm


Still not finding the right product?

Explore all of our products under Polyamide - Nylon 6


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters