Skip to Content
MilliporeSigma

Skip To

GF61487635

Palladium-silver alloy, Pd70Ag30

foil, 50 x 50mm, 0.15mm thickness, as rolled

Synonym(s):

PD070240

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.23
UNSPSC Code:
12352300


form

foil (temper - as rolled)

manufacturer/tradename

Goodfellow 614-876-35

size × thickness

50 mm × 50 mm × 0.15 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters