Skip to Content
MilliporeSigma

Skip To

GF62936062

Polyamide - Nylon 6

(PA 6), rod, diam. 100 mm, L 200 mm, condition oil-filled nylon ′6′

Synonym(s):

AM307950

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.23
UNSPSC Code:
12352100
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

rod

manufacturer/tradename

Goodfellow 629-360-62

L

200 mm

diam.

100 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF92341043GF20583761GF75938721
description

(PA 6), rod, diam. 100 mm, L 200 mm, condition oil-filled nylon ′6′

description

(PA 6), rod, diam. 100 mm, L 200 mm

description

(PA 6), rod, diam. 100 mm, L 1000 mm, condition oil-filled nylon ′6′

description

(PA 6), rod, diam. 100 mm, L 100 mm

diam.

100 mm

diam.

100 mm

diam.

100 mm

diam.

100 mm

L

200 mm

L

200 mm

L

1000 mm

L

100 mm

manufacturer/tradename

Goodfellow 629-360-62

manufacturer/tradename

Goodfellow 923-410-43

manufacturer/tradename

Goodfellow 205-837-61

manufacturer/tradename

Goodfellow 759-387-21

form

rod

form

rod

form

rod

form

rod


Still not finding the right product?

Explore all of our products under Polyamide - Nylon 6


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters