Skip to Content
MilliporeSigma

Skip To

GF74812349

Rhenium

foil, thickness 0.04 mm, length 0.2 m, coil width 0.8 mm

Synonym(s):

RE000229

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
$534.00

About This Item

Empirical Formula (Hill Notation):
Re
CAS Number:
Molecular Weight:
186.21
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141737
MDL number:

$534.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

foil

manufacturer/tradename

Goodfellow 748-123-49

L

0.2 m

coil width

0.8 mm

thickness

0.04 mm

bp

5596 °C (lit.), 5627 °C (lit.)

mp

3180 °C (lit.)

density

21.02 g/cm3 (lit.)

application(s)

catalysts

SMILES string

[ReH]

InChI

1S/Re

InChI key

WUAPFZMCVAUBPE-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF10557288GF50732704GF92641608
description

foil, thickness 0.04 mm, length 0.2 m, coil width 0.8 mm

description

foil, thickness 0.04 mm, length 0.2 m, coil width 0.7 mm

description

foil, thickness 0.04 mm, length 0.5 m, coil width 0.8 mm

description

foil, thickness 0.04 mm, length 0.1 m, coil width 0.8 mm

form

foil

form

foil

form

foil

form

foil

manufacturer/tradename

Goodfellow 748-123-49

manufacturer/tradename

Goodfellow 105-572-88

manufacturer/tradename

Goodfellow 507-327-04

manufacturer/tradename

Goodfellow 926-416-08

density

21.02 g/cm3 (lit.)

density

21.02 g/cm3 (lit.)

density

21.02 g/cm3 (lit.)

density

21.02 g/cm3 (lit.)

mp

3180 °C (lit.)

mp

3180 °C (lit.)

mp

3180 °C (lit.)

mp

3180 °C (lit.)

bp

5596 °C (lit.), 5627 °C (lit.)

bp

5596 °C (lit.), 5627 °C (lit.)

bp

5596 °C (lit.), 5627 °C (lit.)

bp

5596 °C (lit.), 5627 °C (lit.)

application(s)

catalysts

application(s)

catalysts

application(s)

catalysts

application(s)

catalysts


Still not finding the right product?

Explore all of our products under Rhenium


Storage Class

13 - Non Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters