Skip to Content
MilliporeSigma

Skip To

705608

1-Methyluric acid-2,4,5,6-13C4, 1,3,9-15N3

≥98 atom %, ≥98% (CP)

Synonym(s):

1-Methyl-2,6,8-trihydroxypurine-13C4,15N3 (2,4,5,6-13C4, 1,3,9-15N3)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
$1,430.00

About This Item

Empirical Formula (Hill Notation):
13C4C2H615N3NO3
CAS Number:
Molecular Weight:
189.09
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:
Isotopic purity:
≥98 atom %
Assay:
≥98% (CP)
Mass shift:
M+7
Form:
powder

$1,430.00


Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


isotopic purity

≥98 atom %

Quality Segment

assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+7

storage temp.

−20°C

SMILES string

C[15N]1[13C](=O)[15NH][13C]2=[13C](NC(=O)[15NH]2)[13C]1=O

InChI

1S/C6H6N4O3/c1-10-4(11)2-3(9-6(10)13)8-5(12)7-2/h1H3,(H,9,13)(H2,7,8,12)/i2+1,3+1,4+1,6+1,8+1,9+1,10+1

InChI key

QFDRTQONISXGJA-BYSJPOJGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact
Stable Isotopes Customer Service.


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
705616705632705195
description

≥98 atom %, ≥98% (CP)

description

≥98 atom %, ≥98% (CP)

description

≥98 atom %, ≥98% (CP)

description

≥98 atom %, ≥98% (CP)

isotopic purity

≥98 atom %

isotopic purity

≥98 atom %

isotopic purity

≥98 atom %

isotopic purity

≥98 atom %

technique(s)

mass spectrometry (MS): suitable

technique(s)

mass spectrometry (MS): suitable

technique(s)

mass spectrometry (MS): suitable

technique(s)

mass spectrometry (MS): suitable

assay

≥98% (CP)

assay

≥98% (CP)

assay

≥98% (CP)

assay

≥98% (CP)

mass shift

M+7

mass shift

M+7

mass shift

M+7

mass shift

M+7

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters