Skip to Content
MilliporeSigma

Skip To

ADE001099

5-Bromo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine-2-carbaldehyde

AldrichCPR

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Usually ships in 2 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
$1,150.00

About This Item

Empirical Formula (Hill Notation):
C14H9BrN2O3S
Molecular Weight:
365.20
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
NACRES:
NA.21

$1,150.00


Usually ships in 2 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

functional group

aldehyde, bromo

SMILES string

Brc1cnc2n(c(C=O)cc2c1)S(=O)(=O)c3ccccc3

InChI

1S/C14H9BrN2O3S/c15-11-6-10-7-12(9-18)17(14(10)16-8-11)21(19,20)13-4-2-1-3-5-13/h1-9H

InChI key

ZJSMGCHOKHJIFB-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Legal Information

Product of Adesis

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ADVH9C13FFABADE001088AABH9C14B411
description

AldrichCPR

description

-

description

AldrichCPR

description

-

form

solid

form

-

form

solid

form

-

functional group

bromo

functional group

-

functional group

iodo, bromo

functional group

-


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters